Tin ≥99.9985% (metals basis), wire, Puratronic®, Ø 0.5 mm (0.02 in)

Supplier: Thermo Scientific Chemicals

11906-G1 11906-G5 11906-G9
CAAA11906-G1EA 61.15 CAD
CAAA11906-G1 CAAA11906-G5 CAAA11906-G9
Tin ≥99.9985% (metals basis), wire, Puratronic®, Ø 0.5 mm (0.02 in)
Tin

MDL: MFCD00133862


Purity: 99.9985% CAS No.: 7440-31-5 Physical Form: ≈1.43g/m Odor: Odorless MDL No.: MFCD00133862

Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 23-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR