Tin 99,998 Metals basis, wire, Puratronic®, Ø 1.0 mm (0.04 in)
Supplier: Thermo Scientific Chemicals
11905-BW
11905-CB
11905-CD
CAAA11905-BWEA
59.88
CAD
CAAA11905-BW
CAAA11905-CB
CAAA11905-CD
Tin 99,998 Metals basis, wire, Puratronic®, Ø 1.0 mm (0.04 in)
Tin
MDL: MFCD00133862
Purity: 99.998%
CAS No.: 7440-31-5
Physical Form: ≈5.72g/m
Odor: Odorless
MDL No.: MFCD00133862
Formula:
Sn MW: 118.71 g/mol Boiling Pt: 2270 Melting Pt: 232 Density: 7.31 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133862 CAS Number: 7440-31-5 EINECS: 23-141-8 Merck Index: 12,09587 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...