Tin ≥99.9985% (metals basis), wire, Puratronic®, Ø 2.0 mm (0.08 in)

Supplier: Thermo Scientific Chemicals

11904-BU 11904-BY 11904-CC
CAAA11904-BUEA 81.21 CAD
CAAA11904-BU CAAA11904-BY CAAA11904-CC
Tin ≥99.9985% (metals basis), wire, Puratronic®, Ø 2.0 mm (0.08 in)
Tin

MDL: MFCD00133862


Purity: 99.9985% CAS No.: 7440-31-5 Physical Form: ≈22.9g/m Odor: Odorless MDL No.: MFCD00133862

Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 23-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR