Silver diethyldithiocarbamate ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
11898-09
11898-18
CAAA11898-09EA
311.26
CAD
CAAA11898-09
CAAA11898-18
Silver diethyldithiocarbamate ACS
MDL: MFCD00004929
CAS No.: 1470-61-7
Molecular Formula: C5H10AgNS2
Formula Weight: 256.14
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: Yellow to pale brown
Melting Point: 172-175°
MDL No.: MFCD00004929
Formula:
(C₂H₅)₂NCSSAg Melting Pt: 172…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004929 CAS Number: 1470-61-7 EINECS: 216-003-7 UN: 3077 ADR: 9,III |
Specification Test Results
Solubility in pyridine | Pass test |
Suitability for determination of arsenic | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...