Silver benzoate, hydrate
Supplier: Thermo Scientific Chemicals
11896-06
11896-14
11896-22
CAAA11896-06EA
54.67
CAD
CAAA11896-06
CAAA11896-14
CAAA11896-22
Silver benzoate, hydrate
Silver benzoate
MDL: MFCD00013030
CAS No.: 532-31-0
Molecular Formula: C7H5AgO2g.xH2O
Formula Weight: 228.89(anhy)
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: White to cream or grey
MDL No.: MFCD00013030
Formula:
C₇H₅AgO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00013030 CAS Number: 532-31-0 EINECS: 208-533-2 Merck Index: 13,01092 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...