Cadmium ≥99.5% (metals basis), powder -325 mesh

Supplier: Thermo Scientific Chemicals
Danger

11891-09 11891-22 11891-36
CAAA11891-09EA 56.72 CAD
CAAA11891-09 CAAA11891-22 CAAA11891-36
Cadmium ≥99.5% (metals basis), powder -325 mesh
Cadmium

MDL: MFCD00010914


Purity: 99.5% CAS No.: 7440-43-9 Appearance: Silver white UN No.: 3179 MDL No.: MFCD00010914

Formula: Cd
Boiling Pt: 767 °C (1013 hPa)
Melting Pt: 321 °C
Density: 8.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010914
CAS Number: 7440-43-9
EINECS: 231-152-8
UN: 3179
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR