Sodium tetrachloropalladate(II) hydrate (30% Pd) ≥99.95% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Palladium sodium chloride, Sodium palladium (II) chloride, Disodium tetrachloropalladate

11886-04 11886-09 11886-18
CAAA11886-04EA 376.04 CAD
CAAA11886-04 CAAA11886-09 CAAA11886-18
Sodium tetrachloropalladate(II) hydrate (30% Pd) ≥99.95% (metals basis)
Sodium tetrachloropalladate(II) hydrate

MDL: MFCD00003487


Purity: 99.95% CAS No.: 13820-53-6 Molecular Formula: Cl4Na2Pd.xH2O (x≈3) Formula Weight: 294.20(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Red-brown MDL No.: MFCD00003487

Formula: Cl₄Na₂Pd·xH₂O
MW: 294.21 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00003487
CAS Number: 13820-53-6
EINECS: 237-502-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR