Dipotassium tetrachloropalladate ≥99.99% (metals basis) Pd ≥ 32.2%, Premion®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Potassium tetrachloropalladate (II), Potassium chloropalladite

11885-04 11885-09
CAAA11885-04EA 334.96 CAD
CAAA11885-04 CAAA11885-09
Dipotassium tetrachloropalladate ≥99.99% (metals basis) Pd ≥ 32.2%, Premion®
Dipotassium tetrachloropalladate

MDL: MFCD00011373


Purity: 99.99% CAS No.: 10025-98-6 Molecular Formula: K2PdCl4 Formula Weight: 326.42 Physical Form: Crystalline Density: 2.67 Appearance: Red-brown Odor: Odorless Melting Point: 105° dec. MDL No.: MFCD00011373

Formula: Cl₄K₂Pd
MW: 326.43 g/mol
Melting Pt: 105 °C (dec.)
Density: 2.67
Storage Temperature: Ambient
MDL Number: MFCD00011373
CAS Number: 10025-98-6
EINECS: 233-049-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR