Dipotassium hexachloropalladate Pd ≥ 26,3%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Potassium hexachloropalladate(IV)
11884-03
11884-06
CAAA11884-03EA
135.88
CAD
CAAA11884-03
CAAA11884-06
CAAA11884-14
Dipotassium hexachloropalladate Pd ≥ 26,3%
Dipotassium hexachloropalladate
MDL: MFCD00043081
CAS No.: 16919-73-6
Molecular Formula: K2PdCl6
Formula Weight: 397.32
Physical Form: Crystalline
Density: 2.738
Sensitivity: Hygroscopic
Appearance: Red
Odor: Odorless
Melting Point: dec.
UN No.: 3077
MDL No.: MFCD00043081
Formula:
Cl₆K₂Pd MW: 397.33 g/mol Melting Pt: Dec. Density: 2,738 Storage Temperature: Ambient |
MDL Number:
MFCD00043081 CAS Number: 16919-73-6 EINECS: 240-974-6 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...