Dipotassium tetrabromopalladate Pd 20,0%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11883-03
11883-06
CAAA11883-03EA
167.48
CAD
CAAA11883-03
CAAA11883-06
Dipotassium tetrabromopalladate Pd 20,0%
Dipotassium tetrabromopalladate
MDL: MFCD00049659
CAS No.: 13826-93-2
Molecular Formula: K2PdBr4
Formula Weight: 504.21
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Red-brown
Odor: Odorless
MDL No.: MFCD00049659
Formula:
Br₄K₂Pd Storage Temperature: Ambient |
MDL Number:
MFCD00049659 CAS Number: 13826-93-2 EINECS: 237-535-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...