Diammonium tetrachloropalladate Pd ≥ 36%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ammonium tetrachloropalladate(II, Ammonium tetrachloropalladate(II), Ammonium palladium (II) chloride, Ammonium tetrachloropalladate (II)
11882-03
11882-06
CAAA11882-03EA
194.34
CAD
CAAA11882-03
CAAA11882-06
Diammonium tetrachloropalladate Pd ≥ 36%
Diammonium tetrachloropalladate
MDL: MFCD00010884
CAS No.: 13820-40-1
Molecular Formula: H8Cl4N2Pd
Formula Weight: 284.31
Physical Form: Powder
Density: 2.17
Sensitivity: Hygroscopic
Appearance: Dark green to brown
Melting Point: dec.
MDL No.: MFCD00010884
Formula:
H₈Cl₄N₂Pd MW: 284.31 g/mol Melting Pt: Dec. Density: 2,17 Storage Temperature: Ambient |
MDL Number:
MFCD00010884 CAS Number: 13820-40-1 EINECS: 237-498-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...