Palladium(II) iodide (≥28% Pd) ≥99.9% (metals basis) Pd ≥ 28%

Supplier: Thermo Scientific Chemicals
Warning

11881-04 11881-09
CAAA11881-04EA 301.78 CAD
CAAA11881-04 CAAA11881-09
Palladium(II) iodide (≥28% Pd) ≥99.9% (metals basis) Pd ≥ 28%
Palladium (II) iodide

MDL: MFCD00011171


Purity: 99.9% CAS No.: 7790-38-7 Molecular Formula: PdI2 Formula Weight: 360.23 Physical Form: Powder Density: 6.003 Appearance: Black Melting Point: 350° dec. MDL No.: MFCD00011171

Formula: I₂Pd
MW: 360.23 g/mol
Melting Pt: 350 °C (dec.)
Density: 6,003
Storage Temperature: Ambient
MDL Number: MFCD00011171
CAS Number: 7790-38-7
EINECS: 232-203-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR