Palladium(II) bromide ≥99.99% (metals basis) Pd ≥ 39.5%, Premion®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Palladium dibromide, Palladium bromide

11878-04 11878-09
CAAA11878-04EA 417.12 CAD
CAAA11878-04 CAAA11878-09
Palladium(II) bromide ≥99.99% (metals basis) Pd ≥ 39.5%, Premion®
Palladium (II) bromide

MDL: MFCD00011170


Purity: 99.99% CAS No.: 13444-94-5 Molecular Formula: PdBr2 Formula Weight: 266.22 Physical Form: Powder Sensitivity: Hygroscopic MDL No.: MFCD00011170

Formula: Br₂Pd
Melting Pt: Dec.
Density: 5.173 g/cm³ (16 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011170
CAS Number: 13444-94-5
EINECS: 236-588-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR