Silver(I) hexafluorophosphate 98%

Supplier: Thermo Scientific Chemicals
Danger

11873-03 11873-06 11873-14
CAAA11873-03EA 82.48 CAD
CAAA11873-03 CAAA11873-06 CAAA11873-14
Silver(I) hexafluorophosphate 98%
Silver (I) hexafluorophosphate

MDL: MFCD00003415


Purity: 98% CAS No.: 26042-63-7 Molecular Formula: AgF6P Formula Weight: 252.83 Physical Form: Powder Sensitivity: Light Sensitive Appearance: White Odor: Odorless Melting Point: 102° dec. UN No.: 3260 MDL No.: MFCD00003415

Formula: AgF₆P
Melting Pt: 102 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003415
CAS Number: 26042-63-7
EINECS: 247-428-6
UN: 3260
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR