Copper(I) chloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
11871-22
11871-36
11871-A3
CAAA11871-22EA
50.88
CAD
CAAA11871-22
CAAA11871-36
CAAA11871-A3
Copper(I) chloride 97%
Copper (I) chloride
MDL: MFCD00010971
Purity: 97%
CAS No.: 7758-89-6
Molecular Formula: CuCl
Formula Weight: 98.99
Physical Form: Powder
Density: 4.14
Refractive Index: 1.93
Sensitivity: Air and Moisture Sensitive
Appearance: Grey to green
Melting Point: 430°
Boiling Point: 1490°
Merck Reference: 14,2660
UN No.: 2802
MDL No.: MFCD00010971
Formula:
ClCu Boiling Pt: 1490 °C Melting Pt: 430 °C Density: 4.14 Storage Temperature: Ambient |
MDL Number:
MFCD00010971 CAS Number: 7758-89-6 EINECS: 231-842-9 UN: 2802 ADR: 8,III Merck Index: 14,2660 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...