Manganese(II) oxide Mn 76,0-78,0% 99% (76.0 - 78.0% Mn) typ. 99%

Supplier: Thermo Scientific Chemicals
Warning

11870-36 11870-A3 11870-F7
CAAA11870-36EA 77.26 CAD
CAAA11870-36 CAAA11870-A3 CAAA11870-F7
Manganese(II) oxide Mn 76,0-78,0% 99% (76.0 - 78.0% Mn) typ. 99%
Manganese (II) oxide

MDL: MFCD00016226


Purity: 99% CAS No.: 1344-43-0 Molecular Formula: MnO Formula Weight: 70.94 Physical Form: +200 Mesh Powder Density: 5.0 Appearance: Green Odor: Odorless MDL No.: MFCD00016226

Formula: MnO
Melting Pt: 1840 °C
Density: 5,0
Storage Temperature: Ambient
MDL Number: MFCD00016226
CAS Number: 1344-43-0
EINECS: 215-695-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR