Manganese(II) chloride, anhydrous 97%

Supplier: Thermo Scientific Chemicals
Danger

11868-36 11868-A3 11868-A9
CAAA11868-36EA 69.36 CAD
CAAA11868-36 CAAA11868-A3 CAAA11868-A9
Manganese(II) chloride, anhydrous 97%
Manganese (II) chloride

MDL: MFCD00011114


Purity: 97% CAS No.: 7773-01-5 Molecular Formula: Cl2Mn Formula Weight: 125.84 Physical Form: Crystalline/Flake Density: 2.977 Appearance: Pink Odor: Odorless Melting Point: 650° Boiling Point: 1190° Merck Reference: 14,5728 UN No.: 3288 MDL No.: MFCD00011114

Formula: MnCl₂
Boiling Pt: 1190 °C
Melting Pt: 650 °C
Density: 2.977
Storage Temperature: Ambient
MDL Number: MFCD00011114
CAS Number: 7773-01-5
EINECS: 231-869-6
UN: 3288
ADR: 6.1,III
Merck Index: 14,5728

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR