Cadmium acetate dihydrate ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

11865-14 11865-22 11865-36
CAAA11865-14EA 195.92 CAD
CAAA11865-14 CAAA11865-22 CAAA11865-36
Cadmium acetate dihydrate ≥99.999% (metals basis)
Cadmium acetate dihydrate

MDL: MFCD00012449


Purity: 99.999% CAS No.: 5743-04-4 Molecular Formula: Cd(OOCCH3)2.2H2O Formula Weight: 266.54 (230.50anhy) Physical Form: Powder Density: 2.01 Appearance: White Merck Reference: 14,1614 UN No.: 2570 MDL No.: MFCD00012449

Formula: (H₃CCOO)₂Cd·2H₂O
Boiling Pt: 105…107 °C (4 mmHg)
Melting Pt: 256 °C
Density: 2.01
Storage Temperature: Ambient
MDL Number: MFCD00150020
CAS Number: 5743-04-4
EINECS: 208-853-2
UN: 2570
ADR: 6.1,III
Merck Index: 14,1614

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR