Cadmium(II) sulfate, anhydrous 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
11861-14
11861-22
CAAA11861-14EA
51.67
CAD
CAAA11861-14
CAAA11861-22
Cadmium(II) sulfate, anhydrous 99+% ACS
Cadmium (II) sulphate
MDL: MFCD00010923
Purity: 99+%
CAS No.: 10124-36-4
Molecular Formula: CdSO4
Formula Weight: 208.46
Physical Form: Powder
Density: 4.691
Appearance: White
Odor: Odorless
Melting Point: 1000°
Merck Reference: 14,1627
UN No.: 2570
MDL No.: MFCD00010923
Formula:
CdO₄S Melting Pt: 1000 °C Density: 4.691 Storage Temperature: Ambient |
MDL Number:
MFCD00010923 CAS Number: 10124-36-4 EINECS: 233-331-6 UN: 2570 ADR: 6.1,III Merck Index: 14,1627 |
Specification Test Results
Insoluble matter | 0.005% |
Loss on drying at 150°C | 0.01% |
Cl | 0.001% |
Nitrate and nitrite (as NO3) | 0.003% |
Ca | 0.01% |
Cu | 0.002% |
Fe | 0.001% |
PB | 0.003% |
K | 0.02% |
NA | 0.05% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...