Indium(III) chloride, anhydrous ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
11856-03
11856-09
11856-18
11856-30
CAAA11856-03EA
74.58
CAD
CAAA11856-03
CAAA11856-09
CAAA11856-18
CAAA11856-30
Indium(III) chloride, anhydrous ≥99.999% (metals basis)
Indium (III) chloride
MDL: MFCD00011058
Fieser: 16,181 19,173 20,197 21,228
Purity: 99.999%
CAS No.: 10025-82-8
Molecular Formula: InCl3
Formula Weight: 221.18
Physical Form: Crystalline
Density: 3.460
Appearance: White
Odor: Odorless
Melting Point: 586°
Boiling Point: 800°
Merck Reference: 14,4958
UN No.: 3260
MDL No.: MFCD00011058
Formula:
Cl₃In Boiling Pt: 800 °C Melting Pt: 586 °C Density: 3.460 Storage Temperature: Ambient |
MDL Number:
MFCD00011058 CAS Number: 10025-82-8 EINECS: 233-043-0 UN: 3260 ADR: 8,II Merck Index: 14,4958 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...