Phosphorus(V) chloride ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
11849-14
11849-22
CAAA11849-14EA
251.22
CAD
CAAA11849-14
CAAA11849-22
Phosphorus(V) chloride ≥99.99% (metals basis)
Phosphorus (V) chloride
MDL: MFCD00011439
Fieser: 1,866 4,388 5,534 7,290
Purity: 99.99%
CAS No.: 10026-13-8
Molecular Formula: PCl5
Formula Weight: 208.24
Physical Form: Solid
Density: 2.114
Appearance: Yellow
Odor: Pungent
Melting Point: 162° subl.
Merck Reference: 14,7351
UN No.: 1806
MDL No.: MFCD00011439
Formula:
Cl₅P Melting Pt: 162 °C (subl.) Density: 2.114 Storage Temperature: Ambient |
MDL Number:
MFCD00011439 CAS Number: 10026-13-8 EINECS: 233-060-3 UN: 1806 ADR: 8,II Merck Index: 14,7351 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...