Bismuth triacetate ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Acetic acid bismuth (III) salt, Bismuth (III) acetate, Bismuth(III) acetate

11846-09 11846-18
CAAA11846-09EA 178.54 CAD
CAAA11846-09 CAAA11846-18
Bismuth triacetate ≥99.999% (metals basis)
Bismuth triacetate

MDL: MFCD00015629


Purity: 99.999% CAS No.: 22306-37-2 Molecular Formula: Bi(OOCCH3)3 Formula Weight: 386.12 Physical Form: Crystalline Sensitivity: Hygroscopic MDL No.: MFCD00015629

Formula: (H₃CCOO)₃Bi
MW: 386.11 g/mol
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00015629
CAS Number: 22306-37-2
EINECS: 244-904-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR