Thallium(I) acetate ≥99.995% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
11842-14
11842-22
CAAA11842-14EA
372.88
CAD
CAAA11842-14
CAAA11842-22
Thallium(I) acetate ≥99.995% (metals basis)
Thallium (I) acetate
MDL: MFCD00013045
Beilstein Registry No.: 3693914
Soluble in water, alcohol
Purity: 99.995%
CAS No.: 563-68-8
Molecular Formula: Tl(OOCCH3)
Formula Weight: 263.42
Physical Form: Wet Crystalline Aggregates
Sensitivity: Hygroscopic
Solubility: Soluble in water, alcohol
Beilstein Reference: 3693914
Merck Reference: 14,9257
UN No.: 1707
MDL No.: MFCD00013045
Formula:
C₂H₃O₂Tl Boiling Pt: 1457 °C (1013 hPa) Melting Pt: 131 °C Density: 3.68 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013045 CAS Number: 563-68-8 EINECS: 209-257-5 UN: 1707 ADR: 6.1,II Merck Index: 14,9257 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...