Thallium(I) bromide ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Thallium bromide
11841-14
11841-22
CAAA11841-14EA
308.1
CAD
CAAA11841-14
CAAA11841-22
Thallium(I) bromide ≥99.999% (metals basis)
Thallium (I) bromide
MDL: MFCD00011273
Practically insoluble in water
Purity: 99.999%
CAS No.: 7789-40-4
Molecular Formula: TlBr
Formula Weight: 284.28
Physical Form: Powder
Density: 7.5
Melting Point: ca 460°
Solubility: Practically insoluble in water
Merck Reference: 14,9258
UN No.: 1707
MDL No.: MFCD00011273
Formula:
BrTl MW: 284.29 Boiling Pt: 815 Melting Pt: 460 Density: 7.5 Storage Temperature: Ambient |
MDL Number:
MFCD00011273 CAS Number: 7789-40-4 EINECS: 232-163-0 UN: 1707 ADR: 6.1 II Merck Index: 14,9258 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...