Thallium(I) chloride ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
11839-06
11839-14
11839-22
CAAA11839-06EA
103.02
CAD
CAAA11839-06
CAAA11839-14
CAAA11839-22
Thallium(I) chloride ≥99.999% (metals basis)
Thallium (I) chloride
MDL: MFCD00011274
Purity: 99.999%
CAS No.: 7791-12-0
Molecular Formula: TlCl
Formula Weight: 239.82
Physical Form: Powder
Density: 7.0
Refractive Index: 2.247
Sensitivity: Air Sensitive
Melting Point: 430°
Merck Reference: 14,9260
UN No.: 1707
MDL No.: MFCD00011274
Formula:
ClTl Boiling Pt: 720 °C (1013 hPa) Melting Pt: 430 °C Density: 7.0 Storage Temperature: Ambient |
MDL Number:
MFCD00011274 CAS Number: 7791-12-0 EINECS: 232-241-4 UN: 1707 ADR: 6.1,II Merck Index: 14,9260 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...