Hafnium dioxide ≥99.9% (metals basis excluding Zr), Spectrographic Grade (Zr typ. <80 ppm)
Supplier: Thermo Scientific Chemicals
11836-04
11836-09
CAAA11836-04EA
152
CAD
CAAA11836-04
CAAA11836-09
Hafnium dioxide ≥99.9% (metals basis excluding Zr), Spectrographic Grade (Zr typ. <80 ppm)
Hafnium dioxide
MDL: MFCD00003565
Purity: 99.9%
CAS No.: 12055-23-1
Molecular Formula: HfO2
Formula Weight: 210.49
Physical Form: -325 Mesh Powder
Density: 9.68
Melting Point: 2774°
Merck Reference: 14,4588
MDL No.: MFCD00003565
Formula:
HfO₂ Boiling Pt: 5400 °C (1013 hPa) Melting Pt: 2774 °C Density: 9.68 Storage Temperature: Ambient |
MDL Number:
MFCD00003565 CAS Number: 12055-23-1 EINECS: 235-013-2 Merck Index: 14,4588 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...