Hafnium(IV) chloride 98+% (metals basis excluding Zr) (max. 2.7% Zr)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Hafnium tetrachloride
11834-14
11834-22
11834-88
CAAA11834-14EA
111.71
CAD
CAAA11834-14
CAAA11834-22
CAAA11834-88
Hafnium(IV) chloride 98+% (metals basis excluding Zr) (max. 2.7% Zr)
Hafnium (IV) chloride
MDL: MFCD00011034
Notes: Hydrolyzed by water to Hafnyl chloride
Purity: 98+%
CAS No.: 13499-05-3
Molecular Formula: Cl4Hf
Formula Weight: 320.30
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White
Odor: Irritating
Melting Point: 320° subl.
Merck Reference: 14,4588
UN No.: 3260
MDL No.: MFCD00011034
Formula:
Cl₄Hf MW: 320.3 g/mol Melting Pt: 320 °C (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD00011034 CAS Number: 13499-05-3 EINECS: 236-826-5 UN: 3260 ADR: 8,II Merck Index: 14,4588 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...