Molybdenum pentachloride ≥99.6% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

11832-06 11832-18 11832-30 11832-A1
CAAA11832-06EA 72.05 CAD
CAAA11832-06 CAAA11832-18 CAAA11832-30 CAAA11832-A1
Molybdenum pentachloride ≥99.6% (metals basis)
Molybdenum pentachloride

MDL: MFCD00003467


Purity: 99.6% CAS No.: 10241-05-1 Molecular Formula: MoCl5 Formula Weight: 273.24 Physical Form: Crystalline Sensitivity: Moisture Sensitive Appearance: Black Odor: Pungent Melting Point: 194° UN No.: 2508 MDL No.: MFCD00003467

Formula: Cl₅Mo
MW: 273.2 g/mol
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 194 °C
Density: 2,928 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003467
CAS Number: 10241-05-1
EINECS: 233-575-3
UN: 2508
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR