Tungsten trioxide ≥99.8% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
In manufacture of tungsten and tungstates, and in alloys, in fire-proofing fabrics, and as a ceramic pigment
Purity: 99.8%
CAS No.: 1314-35-8
Molecular Formula: O3W
Formula Weight: 231.85
Physical Form: 10-20 Micron Powder
Density: 7.16
Appearance: Yellow
Odor: Odorless
Melting Point: 1473°
Solubility: Insoluble in water and acids. Soluble in hot alkalies. Slightly soluble in HF
Merck Reference: 14,9817
MDL No.: MFCD00011466
In manufacture of tungsten and tungstates, and in alloys, in fire-proofing fabrics, and as a ceramic pigment
Formula:
O₃W Melting Pt: 1473 °C Density: 7.16 Storage Temperature: Ambient |
MDL Number:
MFCD00011466 CAS Number: 1314-35-8 EINECS: 215-231-4 Merck Index: 14,9817 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...