Ruthenium(IV) oxide, anhydrous ≥99.95% (metals basis) Ru ≥ 75.2%, Premion®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ruthenium dioxide, Ruthenium tetroxide
11804-02
11804-04
11804-09
CAAA11804-02EA
83.11
CAD
CAAA11804-02
CAAA11804-04
CAAA11804-09
Ruthenium(IV) oxide, anhydrous ≥99.95% (metals basis) Ru ≥ 75.2%, Premion®
Ruthenium (IV) oxide
MDL: MFCD00011210
Insoluble in water, acids. Soluble in fused alkalis
Purity: 99.95%
CAS No.: 12036-10-1
Molecular Formula: O2Ru
Formula Weight: 133.07
Physical Form: Powder
Density: 6.97
Appearance: Black
Odor: Odorless
Melting Point: 1200° subl.
Solubility: Insoluble in water, acids. Soluble in fused alkalis
UN No.: 1479
MDL No.: MFCD00011210
Formula:
O₂Ru MW: 133.07 g/mol Melting Pt: 1200 °C (subl.) Density: 6.97 Storage Temperature: Ambient |
MDL Number:
MFCD00074857 CAS Number: 12036-10-1 EINECS: 234-840-6 UN: 1479 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...