Ruthenium(IV) oxide hydrate Ru ≥ 54%
Supplier: Thermo Scientific Chemicals
|
Warning
|
11803-03
11803-06
11803-14
CAAA11803-03EA
141.09
CAD
CAAA11803-03
CAAA11803-06
CAAA11803-14
Ruthenium(IV) oxide hydrate Ru ≥ 54%
Ruthenium (IV) oxide hydrate
Oxidation
CAS No.: 32740-79-7
Molecular Formula: O2Ru.xH2O
Formula Weight: 133.07(anhy)
Physical Form: Powder
Appearance: Black
Odor: Odorless
UN No.: 1479
MDL No.: MFCD00149846
Oxidation
Formula:
O₂Ru·H₂O Melting Pt: 1200 °C (subl.) Density: 6.97 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149846 CAS Number: 32740-79-7 EINECS: 234-840-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...