Palladium 5% on barium sulphate, powder hydrogenation catalyst, reduced

Supplier: Thermo Scientific Chemicals

11722-14 11722-22 11722-88
CAAA11722-14EA 654.12 CAD
CAAA11722-14 CAAA11722-22 CAAA11722-88
Palladium 5% on barium sulphate, powder hydrogenation catalyst, reduced
Palladium

MDL: MFCD03613605


Appearance: Grey-black MDL No.: MFCD03613605

Formula: Pd
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR