Antimony (III) sulfide ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Needle antimony, Diantimony trisulfide, Antimonous sulfide, Antimony(III) sulphide, Amtimony sulphide, Diantimony trisulphide, Antimony sulfide black, Stibnite, Antimony glance, Antimony(III) sulfide, Antimony trisulphide, Amtimony sulfide, Antimonous sulphide, Antimony trisulfide, Antimony sulphide black

11683-09 11683-18
CAAA11683-09EA 170.64 CAD
CAAA11683-09 CAAA11683-18
Antimony (III) sulfide ≥99.999% (metals basis)
Antimony (III) sulphide

MDL: MFCD00011217


Purity: 99.999% CAS No.: 1345-04-6 Molecular Formula: Sb2S3 Formula Weight: 339.68 Physical Form: -60 Mesh Powder Melting Point: 550° UN No.: 1549 MDL No.: MFCD00011217

Formula: S₃Sb₂
MW: 339.72 g/mol
Boiling Pt: 1150
Melting Pt: 550
Density: 4.12 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011217
CAS Number: 1345-04-6
EINECS: 215-713-4
UN: 1549
ADR: 6.1 III
Merck Index: 14,713

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR