Arsenic(III) chloride ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
11681-14
11681-22
CAAA11681-14EA
281.24
CAD
CAAA11681-14
CAAA11681-22
Arsenic(III) chloride ≥99.999% (metals basis)
Arsenic (III) chloride
MDL: MFCD00014165
Purity: 99.999%
CAS No.: 7784-34-1
Molecular Formula: AsCl3
Formula Weight: 181.27
Physical Form: Liquid
Density: 2.163
Appearance: Colorless to yellow-brown
Odor: Acrid
Melting Point: -16°
Boiling Point: 130.2°
Merck Reference: 14,801
UN No.: 1560
MDL No.: MFCD00014165
Formula:
AsCl₃ MW: 181.28 Boiling Pt: 130.1 Melting Pt: -8.5 Density: 2.163 Storage Temperature: Ambient |
MDL Number:
MFCD00014165 CAS Number: 7784-34-1 EINECS: 232-059-5 UN: 1560 ADR: 6.1 I Merck Index: 14,801 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...