Ammonium iodide ≥99.0% ACS

Supplier: Thermo Scientific Chemicals
Warning

11668-22 11668-36
CAAA11668-22EA 132.09 CAD
CAAA11668-22 CAAA11668-36
Ammonium iodide ≥99.0% ACS
Ammonium iodide

MDL: MFCD00011424 Freely soluble in water, alcohol, glycerol, methanol


Purity: 99.0% min CAS No.: 12027-06-4 Molecular Formula: NH4I Formula Weight: 144.94 Physical Form: Granular Density: 2.514 Refractive Index: 1.7031 Sensitivity: Light and Air Sensitive and Hygroscopic Appearance: White to pale yellow Odor: Odorless Melting Point: 551° subl. Solubility: Freely soluble in water, alcohol, glycerol, methanol Merck Reference: 14,529 MDL No.: MFCD00011424

Formula: NH₄I
Melting Pt: 551 °C (subl.)
Density: 2.514
Storage Temperature: Ambient
MDL Number: MFCD00011424
CAS Number: 12027-06-4
EINECS: 234-717-7
Merck Index: 14,529

Order Now

Specification Test Results

Insoluble matter 0.005%
Residue after ignition 0.05%
Chloride and bromide (as Cl) 0.005%
PO3 0.00001
SO3 0.05%
Ba 0.002%
Heavy Metals (as Pb) 0.001%
Fe 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR