Sodium sulfide hydrate, tech.

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: di-Sodium sulphide hydrate, di-Sodium sulfide hydrate

11664-36 11664-A3
CAAA11664-36EA 98.91 CAD
CAAA11664-36 CAAA11664-A3
Sodium sulfide hydrate, tech.
Sodium sulphide hydrate

Sodium sulfide hydrate is used as an oxygen scavenger in water treatment, bleaching agent in textile industry, desulfurising and as a dechlorinating agent and in leather industry, sulfonation and sulfomethylation agent in chemical industry.


It is involved in the production of rubber chemicals and sulfur dyes. It is also used to prepare 3-nitroaniline from 1,3-dinitrobenzene and thioethers from alkyl halide. It plays an important role in other applications such as ore flotation, oil recovery, making dyes, and detergent. In addition, it is used in pulp and paper industry in the Kraft process.

Formula: Na₂S·xH₂O
MW: 96.06 g/mol (anhy)
Boiling Pt: ∼920 °C (1013 hPa)
Melting Pt: ∼ 50 °C (dec.)
Density: 1.427
Storage Temperature: Ambient
MDL Number: MFCD00149183
CAS Number: 27610-45-3
EINECS: 215-211-5
UN: 1849
ADR: 8,II
REACH: 01-2119513694-38
Merck Index: 14,8681

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR