Silver(I) acetate, anhydrous 99%

Supplier: Thermo Scientific Chemicals
Warning

11660-14 11660-22
CAAA11660-14EA 211.72 CAD
CAAA11660-14 CAAA11660-22
Silver(I) acetate, anhydrous 99%
Silver (I) acetate

MDL: MFCD00012446 Soluble in dilute HNO3. Water solubility increases with temperature


Purity: 99% CAS No.: 563-63-3 Molecular Formula: C2H3AgO2 Formula Weight: 166.92 Physical Form: Crystalline Density: 3.26 Appearance: White to grey Odor: Odorless Melting Point: dec. Solubility: Soluble in dilute HNO3. Water solubility increases with temperature Merck Reference: 14,8505 UN No.: 3077 MDL No.: MFCD00012446

Formula: C₂H₃AgO₂
Melting Pt: Dec.
Density: 3.26
Storage Temperature: Ambient
MDL Number: MFCD00012446
CAS Number: 563-63-3
EINECS: 209-254-9
UN: 3077
ADR: 9,III
Merck Index: 14,8505

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR