Barium perchlorate trihydrate, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

11656-30 11656-A1
CAAA11656-30EA 93.06 CAD
CAAA11656-30 CAAA11656-A1
Barium perchlorate trihydrate, REacton®, Reagent Grade
Barium perchlorate trihydrate

MDL: MFCD00149148 Soluble in water, methanol. Slightly soluble in ethanol, ethyl acetate, acetone. Insoluble in ether


CAS No.: 10294-39-0 Molecular Formula: Ba(ClO4)2.3H2O Formula Weight: 390.29 (336.24anhy) Physical Form: Crystalline Density: 2.74 Refractive Index: 1.533 Odor: Odorless Solubility: Soluble in water, methanol. Slightly soluble in ethanol, ethyl acetate, acetone. Insoluble in ether Merck Reference: 14,987 UN No.: 1447 MDL No.: MFCD00149148

Formula: BaCl₂O₈·3H₂O
Melting Pt: 70 °C
Density: 2.74
Storage Temperature: Ambient
MDL Number: MFCD00149148
CAS Number: 10294-39-0
EINECS: 236-710-4
UN: 1447
ADR: 5.1,II
Merck Index: 14,987

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR