Copper(II) perchlorate hexahydrate, REacton®, Reagent Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
11651-18
11651-30
CAAA11651-18EA
70.78
CAD
CAAA11651-18
CAAA11651-30
Copper(II) perchlorate hexahydrate, REacton®, Reagent Grade
Copper (II) perchlorate hexahydrate
MDL: MFCD00149666
Soluble in water, methanol, ethanol, acetic acid, acetic anhydride, and acetone
CAS No.: 10294-46-9
Molecular Formula: Cu(ClO4)2.6H2O
Formula Weight: 370.53 (262.44anhy)
Physical Form: Crystalline
Density: 2.23
Refractive Index: 1.505
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 82°
Solubility: Soluble in water, methanol, ethanol, acetic acid, acetic anhydride, and acetone
Merck Reference: 14,2647
UN No.: 1481
MDL No.: MFCD00149666
Formula:
Cl₂CuO₈·6H₂O Melting Pt: 82 °C Density: 2.23 Storage Temperature: Ambient |
MDL Number:
MFCD00149666 CAS Number: 10294-46-9 EINECS: 237-391-4 UN: 1481 ADR: 5.1,II Merck Index: 14,2647 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...