Lanthanum(III) perchlorate hexahydrate ≥99.9%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Lanthanum perchlorate hexahydrate
11641-14
11641-22
CAAA11641-14EA
77.58
CAD
CAAA11641-14
CAAA11641-22
Lanthanum(III) perchlorate hexahydrate ≥99.9%
Lanthanum (III) perchlorate hexahydrate
MDL: MFCD00016159
CAS No.: 14017-46-0
Molecular Formula: La(ClO4)3.6H2O
Formula Weight: 545.35 (437.26anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Odor: Odorless
UN No.: 1481
MDL No.: MFCD00016159
Formula:
Cl₃LaO₁₂·6H₂O MW: 545.35 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00016159 CAS Number: 36907-37-6 EINECS: 237-835-7 UN: 1481 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...