Lithium perchlorate, anhydrous ≥95.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
11639-09
11639-22
11639-36
CAAA11639-09EA
47.72
CAD
CAAA11639-09
CAAA11639-22
CAAA11639-36
Lithium perchlorate, anhydrous ≥95.0% ACS
Lithium perchlorate
As an oxidizing agent
Purity: 95.0% min
CAS No.: 7791-03-9
Molecular Formula: ClLiO4
Formula Weight: 106.40
Physical Form: Crystalline
Density: 2.43
Appearance: White
Melting Point: 236°
Boiling Point: 400° dec.
Solubility: Partially soluble in water (30% at 25°, 72% at 100°). Soluble in alcohol, acetone, ether, ethyl acetate
Merck Reference: 14,5539
UN No.: 1481
MDL No.: MFCD00011079
As an oxidizing agent
Formula:
ClLiO₄ Boiling Pt: 400 °C (dec.) Melting Pt: 236 °C Density: 2.43 Storage Temperature: Ambient |
MDL Number:
MFCD00011079 CAS Number: 7791-03-9 EINECS: 232-237-2 UN: 1481 ADR: 5.1,II Merck Index: 14,5539 |
Specification Test Results
Insoluble matter | 0.005% |
pH of a 5 % solution | 6.0-7.5 at 25° |
Cl | 0.003% |
SO3 | 0.00001 |
Heavy Metals (as Pb) | 5 ppm max. |
Fe | 5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...