Magnesium perchlorate, anhydrous ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Anhydrone
11636-22
11636-36
CAAA11636-22EA
122.13
CAD
CAAA11636-22
CAAA11636-36
Magnesium perchlorate, anhydrous ACS
Magnesium perchlorate
Dessicant. Drying agent for gases
CAS No.: 10034-81-8
Molecular Formula: Mg(ClO4)2
Formula Weight: 223.23
Physical Form: Granular
Density: 2.21
Sensitivity: Hygroscopic
Merck Reference: 14,5678
UN No.: 1475
MDL No.: MFCD00011102
Dessicant. Drying agent for gases
Formula:
Cl₂MgO₈ Melting Pt: 251 °C (dec.) Density: 2.21 Storage Temperature: Ambient |
MDL Number:
MFCD00011102 CAS Number: 10034-81-8 EINECS: 233-108-3 UN: 1475 ADR: 5.1,II Merck Index: 14,5678 |
Specification Test Results
Titratable free acid | 0.005meq/gmeq/g |
Titratable base | 0.025meq/gmeq/g |
Loss on drying at 190 deg C | 8% |
Suitability for moisture absorption | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...