Magnesium perchlorate hexahydrate ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

11635-30 11635-A1
CAAA11635-30EA 131.77 CAD
CAAA11635-30 CAAA11635-A1
Magnesium perchlorate hexahydrate ≥99% (metals basis)
Magnesium perchlorate hexahydrate

MDL: MFCD00149778


Purity: 99% CAS No.: 13446-19-0 Molecular Formula: Mg(ClO4)2.6H2O Formula Weight: 331.33 (223.23anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Merck Reference: 14,5678 UN No.: 1475 MDL No.: MFCD00149778

Formula: Cl₂MgO₈·6H₂O
Melting Pt: 185…190 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149778
CAS Number: 13446-19-0
EINECS: 233-108-3
UN: 1475
ADR: 5.1,II
Merck Index: 14,5678

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR