Nickel(II) perchlorate hexahydrate, REacton®, Reagent Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
11631-14
11631-22
11631-36
CAAA11631-14EA
34.29
CAD
CAAA11631-14
CAAA11631-22
CAAA11631-36
Nickel(II) perchlorate hexahydrate, REacton®, Reagent Grade
Nickel (II) perchlorate hexahydrate
MDL: MFCD00149804
Very soluble in water. Soluble in many organic solvents
CAS No.: 13520-61-1
Molecular Formula: Ni(ClO4)2.6H2O
Formula Weight: 365.70 (257.59anhy)
Physical Form: Damp crystalline solid
Refractive Index: 1.55
Sensitivity: Hygroscopic
Appearance: Green
Odor: Odorless
Melting Point: 140°
Solubility: Very soluble in water. Soluble in many organic solvents
UN No.: 1481
MDL No.: MFCD00149804
Formula:
Cl₂NiO₈·6H₂O Melting Pt: 140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149804 CAS Number: 13520-61-1 EINECS: 237-124-1 UN: 1481 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...