Sodium perchlorate, anhydrous 98.0-102.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
11623-22
11623-36
CAAA11623-22EA
87.06
CAD
CAAA11623-22
CAAA11623-36
Sodium perchlorate, anhydrous 98.0-102.0% ACS
Sodium perchlorate
MDL: MFCD00003480
Purity: 98.0-102.0%
CAS No.: 7601-89-0
Molecular Formula: NaClO4
Formula Weight: 122.46
Physical Form: Crystalline
Density: 2.50
Melting Point: 482° dec.
Merck Reference: 14,8653
UN No.: 1502
MDL No.: MFCD00003480
Formula:
ClNaO₄ Melting Pt: 482 °C (dec.) Density: 2.50 Storage Temperature: Ambient |
MDL Number:
MFCD00003480 CAS Number: 7601-89-0 EINECS: 231-511-9 UN: 1502 ADR: 5.1,II Merck Index: 14,8653 |
Specification Test Results
Insoluble matter | 0.005% |
pH of a 5 % solution | 6.0-8.0 at 25° |
Cl | 0.003% |
SO3 | 0.002% |
Ca | 0.02% |
Heavy Metals (as Pb) | 5 ppm |
Fe | 5 ppm max. |
K | 0.05% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...