Sodium perchlorate monohydrate ≥85.0-90.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Perchloric acid sodium salt
11622-30
11622-A1
CAAA11622-30EA
108.39
CAD
CAAA11622-30
CAAA11622-A1
Sodium perchlorate monohydrate ≥85.0-90.0% ACS
Sodium perchlorate monohydrate
MDL: MFCD00149164
Very soluble in water
Purity: 85.0-90.0%
CAS No.: 7791-07-3
Molecular Formula: NaClO4.H2O
Formula Weight: 140.48 (122.46anhy)
Physical Form: Crystalline
Density: 2.02
Appearance: White
Odor: Odorless
Melting Point: 130° dec.
Solubility: Very soluble in water
Merck Reference: 14,8653
UN No.: 1502
MDL No.: MFCD00149164
Formula:
NaClO₄·H₂O Melting Pt: 130 °C (dec.) Density: 2.02 Storage Temperature: Ambient |
MDL Number:
MFCD00149164 CAS Number: 7791-07-3 EINECS: 231-511-9 UN: 1502 ADR: 5.1,II Merck Index: 14,8653 |
Specification Test Results
Insoluble matter | 0.005% |
pH of a 5% solution | 6.0-8.0 at 25°C |
Cl | 0.003% |
SO3 | 0.002% |
Ca | 0.02% |
K | 0.05% |
Heavy Metals (as Pb) | 5 ppm |
Fe | 5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...