Zinc perchlorate hexahydrate, REacton®, Reagent Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
11614-22
11614-36
CAAA11614-22EA
52.61
CAD
CAAA11614-22
CAAA11614-36
Zinc perchlorate hexahydrate, REacton®, Reagent Grade
Zinc perchlorate hexahydrate
MDL: MFCD00150370
Soluble in water, alcohol
CAS No.: 10025-64-6
Molecular Formula: Cl2O8Zn.6H2O
Formula Weight: 372.37 (264.28anhy)
Physical Form: Crystalline
Density: 2.25
Refractive Index: 1.50
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 106°
Solubility: Soluble in water, alcohol
Merck Reference: 14,10148
UN No.: 1481
MDL No.: MFCD00150370
Formula:
Cl₂O₈Zn·6H₂O Melting Pt: 106 °C Density: 2.25 Storage Temperature: Ambient |
MDL Number:
MFCD00150370 CAS Number: 10025-64-6 EINECS: 237-122-0 UN: 1481 ADR: 5.1,II Merck Index: 14,10148 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...