Silver(II) fluoride 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11610-04
11610-09
11610-18
CAAA11610-04EA
74.89
CAD
CAAA11610-04
CAAA11610-09
CAAA11610-18
Silver(II) fluoride 98+%
Silver (II) fluoride
MDL: MFCD00003411
Purity: 98+%
CAS No.: 7783-95-1
Molecular Formula: AgF2
Formula Weight: 145.87
Physical Form: Crystalline
Density: 4.6
Appearance: Brown
Melting Point: 690°
Merck Reference: 14,8513
UN No.: 3084
MDL No.: MFCD00003411
Formula:
AgF₂ Melting Pt: 690 °C Density: 4.6 Storage Temperature: Ambient |
MDL Number:
MFCD00003411 CAS Number: 7783-95-1 EINECS: 232-037-5 UN: 3084 ADR: 8,II Merck Index: 14,8513 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...