Silver fluoride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11609-06
11609-14
CAAA11609-06EA
136.35
CAD
CAAA11609-06
CAAA11609-14
Silver fluoride 98%
Silver fluoride
MDL: MFCD00003410
Soluble in HF, ammonia, CH3CN
Purity: 98%
CAS No.: 7775-41-9
Molecular Formula: AgF
Formula Weight: 126.87
Physical Form: Crystalline
Density: 5.852
Appearance: Orange/brown
Melting Point: 435°
Boiling Point: ca 1150°
Solubility: Soluble in HF, ammonia, CH3CN
Merck Reference: 14,8514
UN No.: 3260
MDL No.: MFCD00003410
Formula:
AgF Boiling Pt: ∼1150 °C Melting Pt: 435 °C Density: 5.852 Storage Temperature: Ambient |
MDL Number:
MFCD00003410 CAS Number: 7775-41-9 EINECS: 231-895-8 UN: 3260 ADR: 8,II Merck Index: 14,8514 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...