Copper(I) iodide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cuprous iodide

11606-30 11606-A1
CAAA11606-30EA 159.58 CAD
CAAA11606-30 CAAA11606-A1
Copper(I) iodide 98%
Copper (I) iodide

As a catalyst in organic synthesis, cathode ray tube coating, temperature indicator with HgI2


Purity: 98% CAS No.: 7681-65-4 Molecular Formula: CuI Formula Weight: 190.45 Physical Form: Powder Density: 5.63 Refractive Index: 2.346 Sensitivity: Light, Air and Moisture Sensitive Appearance: Pale brown Odor: Weak Melting Point: 605° Boiling Point: ca 1290° Solubility: Insoluble in water, dilute acids, alcohol. Decomposes by conc. acids. Soluble in NH3 solution, iodides, thiosulfates Merck Reference: 14,2662 UN No.: 3077 MDL No.: MFCD00010978


As a catalyst in organic synthesis, cathode ray tube coating, temperature indicator with HgI2

Formula: CuI
MW: 190.45 g/mol
Boiling Pt: 1290
Melting Pt: 602
Density: 5.63
Storage Temperature: Ambient
MDL Number: MFCD00010978
CAS Number: 7681-65-4
EINECS: 231-674-6
UN: 3077
ADR: 9 III
Merck Index: 14,2662

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR